Learn More
Zymo Research Corporation Anti-5-Hydroxymethylcytosine Polyclonal Antibody (25 µg) 1 µg /µl
SDP
Supplier:  Zymo Research Corporation A400125
Encompass_Preferred
Specificity of the antibody is enhanced such that crossreactivity with unmodified and methylated templates is suppressed to near-background levels. The antibody has been extensively tested and validated in ELISA and immunoprecipitation-based enrichment assays, and is suitable for use in further applications including immunohistochemical labeling and chromatographic blotting.
Catalog No. 50-125-1271