Learn More
Zymo Research Corporation Anti-5-Hydroxymethylcytosine Polyclonal Antibody (200 µg) 1 µg /µl
SDP
Supplier:  Zymo Research Corporation A4001200
Encompass_Preferred
The rabbit Anti-5-hmC Polyclonal Antibody can robustly distinguish between hydroxymethylated DNA and methylated or unmodified DNA with limited to no cross-reactivity. The antibody has been validated in ELISA and immunoprecipitation-based enrichment assays, and is suitable for use in further applications including immunohistochemical labeling and chromatographic blotting.
Catalog No. 50-203-6987