Learn More
Zymo Research Corporation Anti-5-Methlycytosine 50 ul @ 1 ug/ul (Clone 7D21)
SDP
Supplier:  Zymo Research Corporation A300250
Encompass_Preferred
The mouse Anti-5-Methylcytosine monoclonal antibody (clone 7D21) has been developed to facilitate differentiation between methylated and non-methylated cytosines in DNA. Specificity of this clone is to 5-methylcytosines in single-stranded DNA with no detectable crossreactivity to non-methylated or hydroxymethylated cytosines. The antibody has proven to be a valuable tool in the characterization of DNA methylation and has been successfully used for immunopreciptiationbased assays, ELISAs and immunofluorescence assays.
Catalog No. 50-195-7570