Learn More
Ubpbio ANTI-6XHIS, MOUSE 100UG
Supplier:  Ubpbio Y1011
Encompass_Preferred
Antibodies – Anti-6XHis (UBPBio catalog# Y1011) – Mouse monoclonal antibody (clone His.H8, IgG2b). Used for immunoblotting at 1:1000-3000 dilution for detection of 1-2 ng His-tagged proteins; immunostaining at 1:500-2,000 dilution.
Catalog No. 50-112-6916