Learn More
AdipoGen anti-8-Hydroxy-2'-deoxyguanosine [8-OHdG], mAb (N45.1)
SDP
Catalog No. 501015750
Encompass_Preferred
anti-8-Hydroxy-2'-deoxyguanosine [8-OHdG], mAb (N45.1), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes 8-OHdG. Does not cross-react with 19 analogues of 8-OHdG (guanosine (G), 7-methylG, 6-SHG, 8-bromoG, dA, dC, dT, dI, dU, dG, O6-methyldG, 8-OHdA, guanine (Gua), O6-methylGua, 8-OHGua, uric acid, urea, creatine, creatinine). Minimal cross-reactivity with 8-sulfhydrylG and 8-OHG (less than 1%). Lyophilized. Contains 10mM PBS, pH7.4 containing 1.0% BSA. 8-hydroxy-2'-deoxyguanosine (8-OHdG) is a modified base that occurs in DNA due to attack by hydroxyl radicals (incl. singlet oxygen and direct photodynamic action) that are formed as byproducts and intermediates of aerobic metabolism and during oxidative stress.
Catalog No. 50-101-5750 Supplier AdipoGen Supplier No. JAIMOG100P
anti-8-Hydroxy-2'-deoxyguanosine [8-OHdG], mAb (N45.1), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes 8-OHdG. Does not cross-react with 19 analogues of 8-OHdG (guanosine (G), 7-methylG, 6-SHG, 8-bromoG, dA, dC, dT, dI, dU, dG, O6-methyldG, 8-OHdA, guanine (Gua), O6-methylGua, 8-OHGua, uric acid, urea, creatine, creatinine). Minimal cross-reactivity with 8-sulfhydrylG and 8-OHG (less than 1%). Lyophilized. Contains 10mM PBS, pH7.4 containing 1.0% BSA. 8-hydroxy-2'-deoxyguanosine (8-OHdG) is a modified base that occurs in DNA due to attack by hydroxyl radicals (incl. singlet oxygen and direct photodynamic action) that are formed as byproducts and intermediates of aerobic metabolism and during oxidative stress.