Learn More
Aviva Systems Biology Corp A630025C20RIK Antibody - N-terminal region (ARP37513_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37513_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against A630025C20RIK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse A630025C20RIK. Alternate names: Mlr2, mKIAA1795, 3110023F06Rik, A630025C20RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53078