Learn More
Aviva Systems Biology Corp A830039H10RIK Antibody - C-terminal region (ARP37512_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37512_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against A830039H10RIK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Mlr1, A830039H10RIKApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53820