Learn More
Medchemexpress LLC Anti-AA2AR Adenosine 5mg | 5mg
SDP
Catalog No. 5000325777
Encompass_Preferred
Anti-AA2AR/Adenosine A2aR Antibody (3F6-9G5) is a humanized antibody expressed in CHO cells targeting AA2AR/Adenosine A2aR Anti-AA2AR/Adenosine A2aR Antibody (3F6-9G5) contains huIgG1 heavy chain and hu light chain with a predicted molecular weight (MW) of 146 66 kDa The isotype control for Anti-AA2AR/Adenosine A2aR Antibody (3F6-9G5) can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-25777 Supplier Medchemexpress LLC Supplier No. HYP9903245MG
Anti-AA2AR/Adenosine A2aR Antibody (3F6-9G5) is a humanized antibody expressed in CHO cells targeting AA2AR/Adenosine A2aR Anti-AA2AR/Adenosine A2aR Antibody (3F6-9G5) contains huIgG1 heavy chain and hu light chain with a predicted molecular weight (MW) of 146 66 kDa The isotype control for Anti-AA2AR/Adenosine A2aR Antibody (3F6-9G5) can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)