Learn More
American Research Products Inc Anti-AAV VP1/VP2/VP3 Monoclonal Antibody, Mouse Clone B1, Robust Detection of AAV Proteins, 5 mL
Supplier:  American Research Products Inc 65158
Encompass
Anti-AAV VP1/VP2/VP3 Monoclonal Antibody, B1 Clone is a versatile tool for detecting AAV capsid proteins, enhancing gene therapy and virology research. It specifically targets free VP1, VP2, and VP3 proteins across various AAV serotypes, differentiating it from similar products. This antibody is optimized for applications including Immunofluorescence and Western Blotting, making it essential for robust characterization of viral infection and vector development.
Catalog No. NC2876414