Learn More
Sino Biological Anti-ACE2 Antibody, Rabbit Polyclonal 20 µL
Catalog No. 5000785902
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat ACE2 (rR ACE2; Catalog#80031-R08H; Q5EGZ1; Met 1-Thr 740). ACE2 specific IgG was purified by Rat ACE2 affinity chromatography.
Catalog No. 50-007-85902 Supplier Sino Biological Supplier No. 80031RP0220
Produced in rabbits immunized with purified, recombinant Rat ACE2 (rR ACE2; Catalog#80031-R08H; Q5EGZ1; Met 1-Thr 740). ACE2 specific IgG was purified by Rat ACE2 affinity chromatography.