Learn More
Genetex Inc Anti-beta Actin antibody [GT5512]
SDP
Supplier:  Genetex Inc GTX629630
Encompass
The Anti-beta Actin antibody [GT5512] from GeneTex is a Mouse Monoclonal antibody to beta Actin. The Anti-beta Actin antibody [GT5512] Recognizes Human, Mouse, Drosophila, Rat, Zebrafish, Yeast, Goat, Rabbit. The Anti-beta Actin antibody [GT5512] has been validated for the following Applications: ICC/IF, IHC-P, WB.
Catalog No. NC1080972