Learn More
ECM Bioscience Actin (Tyr-53), phospho-specific Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience AP1671
Encompass_Preferred
This antibody detects a 42 kDa* protein corresponding to the molecular mass of Actin on SDS-PAGE immunoblots of pervanadate treated human C2C12 and SYF cSrc transformed cells, but not in control cells. In addition, this antibody either detects actin cleavage products at 36 and 20 kDa, or cross-reacts with unidentified proteins at these molecular weights in C2C12 cells. The immunogen was Phospho-Actin (Tyr-53) synthetic peptide (coupled to KLH) corresponding to amino acid residues around tyrosine 53 of human β actin. This sequence is identical to similar regions in all four α actins, as well as in γ actin, and is well conserved in actins from most eukaryotic species. Accession #P60709. Applications = WB, E, ICC; Species Reactivity = Hu, Rt, Ms, Ck; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Actin
Catalog No. 50-206-8615