Learn More
IBL America Anti-Adalimumab (Humira) Confirmatory Reagent
Supplier:  IBL America TM09009CR
Encompass_Preferred
The Confirmatory Reagent contains the drug Infliximab (Remicade) lyophilized. The reconstituted Confirmatory Reagent is designed for use as a competition/inhibition test for the confirmation of samples which have been ADA positive in the initial screening test coded Cat#. Used as a complementary component for Anti-Infliximab (Remicade) (ATI) ELISA.
Catalog No. 50-136-6623