Learn More
BIOHIPPO Anti-Agrobacterium sp. aroA/EPSPS Monoclonal Antibody (1A079), 100 µg
SDP
Catalog No. 5000857983
Encompass_Preferred
Agrobacterium sp Agrobacterium sp. aroA/EPSPS Monoclonal Antibody is a Mouse-derived monoclonal antibody that specifically recognizes Agrobacterium sp. aroA/EPSPS. It shows reactivity with Agrobacterium sp. Validated applications include ELISA, IHC, WB. Protein A/G purified from cell culture supernatant. Supplied as 100 ug.
Catalog No. 50-008-57983 Supplier BIOHIPPO Supplier No. BHA21416298
Agrobacterium sp Agrobacterium sp. aroA/EPSPS Monoclonal Antibody is a Mouse-derived monoclonal antibody that specifically recognizes Agrobacterium sp. aroA/EPSPS. It shows reactivity with Agrobacterium sp. Validated applications include ELISA, IHC, WB. Protein A/G purified from cell culture supernatant. Supplied as 100 ug.