Learn More
ECM Bioscience Akt (N-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience AM1011
Encompass_Preferred
Clone M101 was generated from a recombinant protein containing amino acid residues in the N-terminal region of human Akt1. This sequence is highly conserved in human and mouse Akt, and may recognize Akt2 and Akt3. Accession #P31749. This antibody detects a 60 kDa* protein corresponding to the apparent molecular mass of Akt on SDS-PAGE immunoblots of A431 cell lysate. Similar results were seen in various other human, rat, and mouse lysates. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = PKBalpha, PKB, AKT
Catalog No. 50-206-8608