Learn More
ECM Bioscience ALDH1A1 (N-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience AM5581
Encompass_Preferred
Clone M558 was generated from a recombinant protein corresponding to amino acids in the N-terminal region from human ALDH1A1. This sequence is highly conserved in rat and mouse ALDH1A1. Accession #P00352. The antibody was purified by Protein G chromatography. This antibody antibody detects a 55 kDa* protein on SDS-PAGE immunoblots of human A431, HepG2, and mouse liver, as well as human recombinant ALDH1A1. The antibody does not detect ALDH1A3 in PC3 cells, ALDH1A2 in Jurkat cells, and bovine tubulin (55 kDa). Applications: WB, E; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal purified with protein G chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = RALDH, ALDH-E1, ALHDII, Aldehyde dehydrogenase, ALDC, PUMB1, ALDH1A1
Catalog No. 50-206-8613