Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD anti-Allophycocyanin (APC) Magnetic Particles - DM IMag-DM, Clone: E30-221

Catalog No. BDB557932 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557932 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557932 Supplier BD Supplier No. 557932
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell Separation

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store undiluted at 4°C.
Quantity 5 mL
For Use With (Application) Cell separation
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.