Learn More
Abcam Rabbit recombinant monoclonal antibody, anti-alpha/beta-synuclein, clone EP1646Y, carrier-free, PBS buffer, 100 μg
Supplier: Abcam AB189217100UG
This rabbit recombinant monoclonal antibody, clone EP1646Y, targets both alpha- and beta-synuclein, which are proteins critically involved in neurodegenerative disorders like Parkinson's disease. It is supplied in a carrier-free PBS-only buffer, making it suitable for direct conjugation. The antibody is validated for various applications, including Western Blot and Flow Cytometry, providing a valuable tool for research into synaptic function and related pathologies.
- Targets alpha- and beta-synuclein
- Recombinant monoclonal antibody
- Clone EP1646Y
- Host species is rabbit
- Reactivity with human, mouse, and rat samples
- Applications: Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
- Predicted molecular weight: 14 kDa
- Formulated in PBS-only buffer
- Carrier-free (BSA and Azide free)
- Specific for alpha- and beta-synuclein
- UniProt ID: P37840 (for Alpha-Synuclein)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.