Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Anti-alpha Actinin 4 antibody, mouse monoclonal, 100 kDa, 100 μL, liquid

Supplier:  Abcam AB254074100UL

Encompass_Preferred

This mouse monoclonal Anti-alpha Actinin 4 (ACTN4) antibody [clone 93] is specifically designed to detect the ~100 kDa ACTN4 protein, integral to the cytoskeleton's structure and various cellular processes. It is an effective tool for research on cell motility, adhesion, and signaling pathways. This antibody is validated for Western Blot applications and shows reactivity with mouse samples.

  • Target is Alpha Actinin 4 (ACTN4)
  • Host species is Mouse
  • Monoclonal antibody
  • Validated for Western Blot (WB) applications
  • Target molecular weight is approximately 100 kDa
  • Purified via Protein G affinity
  • Supplied in a buffered liquid formulation
  • Storage buffer includes pH 7.5, HEPES, 50% glycerol
  • Shipping conditions: Blue Ice
  • Short-term storage at 1-2 weeks at +4°C
  • Long-term storage at -20°C
  • Avoid freeze/thaw cycle

Catalog No. NC2471035


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.