Learn More
Abcam Mouse monoclonal [34.2] to AMPK alpha 1 + AMPK alpha 2Suitable for: WBReacts with: Rat, Human, Drosophila melanogasterIsotype: IgG2a
Supplier:  Abcam AB8003950UG
Encompass_Preferred
Mouse monoclonal [34.2] to AMPK alpha 1 + AMPK alpha 2Suitable for: WBReacts with: Rat, Human, Drosophila melanogasterIsotype: IgG2a
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2741289