Learn More
Sino Biological Anti-Annexin V/ANXA5 Antibody, Mouse Monoclonal 100 µL
Catalog No. 5000749521
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Annexin V/ANXA5 (rh Annexin V/ANXA5; Catalog#10448-HNAE; P08758; Met1-Asp320). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-007-49521 Supplier Sino Biological Supplier No. 10448MM03100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Annexin V/ANXA5 (rh Annexin V/ANXA5; Catalog#10448-HNAE; P08758; Met1-Asp320). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.