Learn More
Precision Antibody ANTI-AR-V7 MONOCLN ANTIBODY
Supplier:  Precision Antibody AG10008
Encompass
Murine Monoclonal Anti-Ar-V7. Ihc: Anti-Ar-V7 Recognizes The Androgen Receptor Variant (Ar-V7); But Does Not Cross React With Intact Androgen Receptor (Ar).
Catalog No. NC0752138