Learn More
FUJIFILM Biosciences Anti Arg1 Rabbit 100uL
Catalog No. 5000450172
Encompass_Preferred
Arginase 1 Arg1 is a hydrolase converting arginine into ornithine and urea expressed highly in M2-type microglia/macrophages This product is a rabbit polyclonal antibody that recognizes Arg1 Useful for visualizing Arg1 in immunocytochemistry studies Specific to Arginase 1 Arg1
Catalog No. 50-004-50172 Supplier FUJIFILM Biosciences Supplier No. 01828361
Arginase 1 Arg1 is a hydrolase converting arginine into ornithine and urea expressed highly in M2-type microglia/macrophages This product is a rabbit polyclonal antibody that recognizes Arg1 Useful for visualizing Arg1 in immunocytochemistry studies Specific to Arginase 1 Arg1