Learn More
ECM Bioscience Arp3 (C-terminal region) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience AP4581
Encompass_Preferred
This antibody was affinity purified using Arp3 (C-terminal region) peptide (without carrier). The antibody detects a 47 kDa* doublet corresponding to the molecular mass of Arp3 on SDS-PAGE immunoblots of human A431, Jurkat, and HeLa cells, and in rat PC12 cells. The immunogen was Arp3 (C-terminal region) synthetic peptide (coupled to KLH) corresponding to amino acid residues in the C-terminal region of human Arp3. This peptide sequence is highly conserved in rat, mouse, chicken, fish, and yeast Arp3 proteins. Accession #P61158. Applications = WB, E, ICC, IHC; Species Reactivity = Hu, Rt, Ms, Ck, F; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = ACTR3, Arp3
Catalog No. 50-206-8616