Learn More
AdipoGen anti-Asc, pAb (AL177),Polyclonal Antibody
SDP
Supplier:  AdipoGen AG25B0006C100
Encompass_Preferred
anti-Asc, pAb (AL177), AdipoGen Life Sciences. Polyclonal Antibody. Recognizes human and mouse Asc. Source: Rabbit. Applications: FUNC, ICC, IHC, IP, WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. Asc promotes caspase-mediated apoptosis. This proapoptotic activity is mediated predominantly through the activation of caspase-9. It is a component of the inflammasome, a protein complex which also includes NLRP3/NALP3, CARD8 and CASP1 and whose function is the activation of proinflammatory caspases.
Catalog No. 50-463-608