Learn More
Immunology Consultants Monoclonal Mouse anti-African Swine Fever Virus (ASFV) P72 Antibody Clone #10E11, 1.0 mg
SDP
Supplier:  Immunology Consultants MASF5A10E11
Encompass_Preferred
This mouse IgG1 kappa antibody is purified using Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with ASFV p72 protein as determined by ELISA and western blotting. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-224-8984