Learn More
FUJIFILM Biosciences Anti asialo GM1 Rabbit 1mL
Catalog No. 5000450132
Encompass_Preferred
The glycolipid asialo ganglioside-GM1 ASGM1 is expressed in T cells infected with viruses and in natural killer NK cells ASGM1 is localized in lipid raft structures in NK and CD8 T cells This product is a rabbit polyclonal antibody that recognizes asialo GM1 It is effective in depleting NK cells in various mouse and rat strains making it valuable for immunological research Specific reactivity to asialo GM1
Catalog No. 50-004-50132 Supplier FUJIFILM Biosciences Supplier No. 01409801
The glycolipid asialo ganglioside-GM1 ASGM1 is expressed in T cells infected with viruses and in natural killer NK cells ASGM1 is localized in lipid raft structures in NK and CD8 T cells This product is a rabbit polyclonal antibody that recognizes asialo GM1 It is effective in depleting NK cells in various mouse and rat strains making it valuable for immunological research Specific reactivity to asialo GM1