Learn More
ABclonal Technology ATRNL1 Polyclonal Antibody,100UL,Validated in: WB, IHC
SDP
Supplier:  ABclonal Technology A14868
Encompass_Preferred
Immunogen: Recombinant fusion protein containing a sequence corresponding to amino acids 53-160 of human ATRNL1 (NP_001263211.1).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse•Validated Applications:WB,IHC•Swiss Prot: Q5VV63•Gene ID:26033•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 50-164-9188