Learn More
Biolegend Anti-β-Amyloid, 1-16
Supplier:  Biolegend 803017
Encompass_Preferred
Anti-β-Amyloid, 1-16 [6E10]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, Direct ELISA, IHC-P, IHC-F, EM; Size: 1 mL
Catalog No. 50-208-9884