Learn More
Seracare Life Sciences Inc ANTI-B BURGDORFERI AB FITC
Supplier:  Seracare Life Sciences Inc 53300064
Encompass_Preferred
Affinity purified polyclonal antibody to Borrelia burgdorferi made in Goat and labeled with fluorescein isothiocyanate (FITC). Isolated from a serum pool of goats immunized with heat killed whole cells of Borrelia burgdorferi.
Catalog No. 50-669-23