Learn More
Seracare Life Sciences Inc ANTI-B BURGDORFERI AB RESAP
Supplier:  Seracare Life Sciences Inc 53200048
Encompass_Preferred
Affinity purified polyclonal antibody to Borrelia burgdorferi made in Goat and labeled with alkaline phosphatase. ReserveAP conjugates represent the latest advances in KPL's conjugation chemistry and are more sensitive than KPL's standard AP conjugates. The antibody is highly specific for Borrelia burgdorferi. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-670-16