Learn More
Seracare Life Sciences Inc ANTI-B BURGDORFERI ANTIBODY
Supplier:  Seracare Life Sciences Inc 53100337
Encompass_Preferred
Affinity purified polyclonal antibody to Borrelia burgdorferi made in Goat. Isolated from a serum pool of goats immunized with heat killed whole cells of Borrelia burgdorferi. The antibody is highly specific for Borrelia burgdorferi.
Catalog No. 50-668-68