Learn More
ECM Bioscience β-Catenin (N-terminal) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience CP1061
Encompass_Preferred
This antibody detects a 92kDa* protein corresponding to the molecular mass of β-Catenin on SDS-PAGE immunoblots of Hct116 src transformed cells. Similar results were seen in human endothelial, A431, and HeLa cells. The immunogen was β-Catenin synthetic peptide (coupled to KLH) corresponding to amino acid residues in the N-terminal region of human β-Catenin. This human sequence is highly conserved in rat and mouse β-Catenins. Accession #P35222. Applications = WB, E, IP, IHC; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Catenin beta1, CTNNB1, catenin
Catalog No. 50-206-8674