Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse B Lymphocyte Enrichment Set - DM, BD IMag-DM, Biotin, BD

Catalog No. BDB557792 Shop All BD Biosciences Products
Change view
Click to view available options
Conjugate:
BD IMag-DM, Biotin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
BDB557792 BD IMag-DM, Biotin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557792 Supplier BD Biosciences Supplier No. 557792
Add to Cart
Add to Cart

Cell Separation

TRUSTED_SUSTAINABILITY

Specifications

Classification Monoclonal
Conjugate BD IMag-DM, Biotin
Regulatory Status RUO
Primary or Secondary Primary
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.