Learn More
ECM Bioscience B-Raf (N-terminus) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience RP2011
Encompass_Preferred
This antibody was affinity purified using B-Raf (N-terminus) peptide (without carrier). The antibody detects 105 and 95 kDa* bands corresponding to the molecular mass of B-Raf. The doublet observed in Jurkat cells may represent multipe isoforms of the B-Raf, since all bands are blocked specifically with B-Raf (N-terminus) peptide. The immunogen was A synthetic peptide (coupled to KLH) corresponding to amino acid residues in the N-terminus of human B-Raf. This sequence has high homology with similar regions in rat and mouse B-Raf, and has low homology to other Raf family members. Accession #P15056. Applications = WB, E; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Braf
Catalog No. 50-206-8819