Learn More
ECM Bioscience β-Tubulin (a.a. 168-177) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience TP1781
Encompass_Preferred
This antibody was affinity purified using βIII-Tubulin (a.a. 168-177) peptide (without carrier). This antibody detects a 50 kDa* protein corresponding to the molecular mass of β-Tubulin on SDS-PAGE immunoblots of purified brain tubulin and mouse brain tissue. The immunogen was βIII-Tubulin (a.a. 168-177) synthetic peptide (coupled to KLH) corresponding to amino acid residues from human βIII-Tubulin. This sequence is identical to similar regions in βI, βII, and βIII-Tubulin isotypes, and is well conserved in tubulins from most eukaryotic species. Accession #Q13509. Applications = WB, E, ICC; Species Reactivity = Hu, Rt, Ms, Ck; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Tubulin
Catalog No. 50-206-8865