Learn More
Sino Biological Anti-B7-2/CD86 Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000786596
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CD86/B7-2 (rR CD86/B7-2; Catalog#80025-R08H; NP_064466.1; Met1-Ile250). CD86/B7-2 specific IgG was purified by Rat CD86/B7-2 affinity chromatography.
Catalog No. 50-007-86596 Supplier Sino Biological Supplier No. 80025T16100
Produced in rabbits immunized with purified, recombinant Rat CD86/B7-2 (rR CD86/B7-2; Catalog#80025-R08H; NP_064466.1; Met1-Ile250). CD86/B7-2 specific IgG was purified by Rat CD86/B7-2 affinity chromatography.