Learn More
Sigma Aldrich Fine Chemicals Biosciences Monoclonal Anti-beta-Actin antibody produced in mouse, 100 uL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A5441100UL
Encompass
Monoclonal Anti-beta-Actin antibody produced in mouse has also been used: in immunofluorescence staining, in immunoblotting, in immunohistochemistry, and as a control for protein arrays.
Catalog No. NC0793751