Learn More
Eagle Biosciences Inc ANTI BETA 2 GLYCOPROTEIN 1 SCREEN ELISA
SDP
Supplier:  Eagle Biosciences Inc DKO145
Encompass
Eagle Biosciences Glycoprotein 1 Screen ELISA Assay Kit is an indirect enzyme-linked immunosorbent assay (ELISA) kit designed for the quantitative measurement of IgG or IgM class antibodies directed against the 2-Glycoprotein 1 in human serum or plasma. Anti beta 2 Glycoprotein 1 Screen ELISA Assay Kit is intended for research use only and is not intended for diagnostic procedures.
Catalog No. 50-187-8283