Learn More
Sino Biological Anti-beta Galactosidase Antibody 20 µL
Catalog No. 5000786122
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant E. coli lacZ/beta Galactosidase. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-007-86122 Supplier Sino Biological Supplier No. 69078MM3920
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant E. coli lacZ/beta Galactosidase. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.