Learn More
Sino Biological Anti-beta Galactosidase Antibody, Rabbit Polyclonal 20 µL
Catalog No. 5000786123
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant E. coli lacZ/beta Galactosidase (WP_000177906.1; 1M-1024K). IgG1 specific IgG was purified by E. coli lacZ/beta Galactosidase affinity chromatography.
Catalog No. 50-007-86123 Supplier Sino Biological Supplier No. 69078T1620
Produced in rabbits immunized with purified, recombinant E. coli lacZ/beta Galactosidase (WP_000177906.1; 1M-1024K). IgG1 specific IgG was purified by E. coli lacZ/beta Galactosidase affinity chromatography.