Learn More
STEMCELL Technologies Anti-Beta-Tubulin III Antibody, Clone AA10, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 60100
Encompass
The AA10 antibody reacts with beta-tubulin III, an ~50 - 55 kDa structural protein that is a constituent of tubulin. Tubulin is the major component of microtubules within the cytoskeleton and is assembled from heterodimers of alpha and beta tubulin subunits. The beta III isoform of tubulin, also known as neuron-specific class III beta-tubulin, is expressed primarily in neurons and is widely used as a marker to distinguish neurons from other cell types. Beta-tubulin III contributes to microtubule formation in neuronal cell bodies and axons, a function involving GTP binding, and plays roles in axonal transport, neuronal cell proliferation, and differentiation. It is highly expressed in several types of cancer and is a predictive and prognostic marker for various tumors, for example, being found in neoplastic but not in normal glial cells. The AA10 antibody is expected to recognize all mammalian homologs of beta-tubulin III.
Catalog No. 50-197-5544