Learn More
LAB BIOREAGENTS Anti-Bevacizumab Elisa Kit
Supplier:  LAB BIOREAGENTS BEK021132
Encompass
Anti-Bevacizumab ELISA Kit is a sandwich enzyme-linked immunoassay for the quantitative determination of Anti-Bevacizumab with high sensitivity and specificity in serum and plasma.Kit PerformanceSensitivity: 10 ng/mLCross-reactivity: There is no cross reaction with native serum immunoglobulinsPrecision: Intra-Assay: CV < 20%; Inter-Assay: CV < 20%
Catalog No. 50-195-9286