Learn More
Sino Biological Anti-Bikunin/AMBP Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000751299
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Bikunin/AMBP (rh Bikunin/AMBP; Catalog#13141-H08H1; NP_001624.1; Met1-Val203). Bikunin/AMBP specific IgG was purified by Human Bikunin/AMBP affinity chromatography.
Catalog No. 50-007-51299 Supplier Sino Biological Supplier No. 13141T16100
Produced in rabbits immunized with purified, recombinant Human Bikunin/AMBP (rh Bikunin/AMBP; Catalog#13141-H08H1; NP_001624.1; Met1-Val203). Bikunin/AMBP specific IgG was purified by Human Bikunin/AMBP affinity chromatography.