Learn More
Bioworld ANTI-BOVINE ALBUMINBSA) R
Supplier:  Bioworld 201810892
Encompass
ANTI-BOVINE ALBUMIN (BSA) RED SEPAROPORE (AGAROSE): Application: For the purification of Bovine Albumin (BSA). Specifications: Ligand: Anti-BSA Antibody; Particle size (range): 52-165mMatrix: Red Separopore 4B (Agarose Beads, 4%); Ligand...
Catalog No. 50-197-898