Learn More
Bioworld ANTI-BOVINE ALBUMINBSA) R
Catalog No. 50197897
Encompass
ANTI-BOVINE ALBUMIN (BSA) RED SEPAROPORE (AGAROSE): Application: For the purification of Bovine Albumin (BSA). Specifications: Ligand: Anti-BSA Antibody; Particle size (range): 52-165mMatrix: Red Separopore 4B (Agarose Beads, 4%); Ligand...
Catalog No. 50-197-897 Supplier Bioworld Supplier No. 201810891
ANTI-BOVINE ALBUMIN (BSA) RED SEPAROPORE (AGAROSE): Application: For the purification of Bovine Albumin (BSA). Specifications: Ligand: Anti-BSA Antibody; Particle size (range): 52-165mMatrix: Red Separopore 4B (Agarose Beads, 4%); Ligand...