Learn More
Bioworld ANTI-BOVINE ALBUMINBSA) S
Supplier:  Bioworld 201810591
Encompass
ANTI-BOVINE ALBUMIN (BSA) SEPAROPORE (AGAROSE): Application: For the purification of Bovine Albumin (BSA). Specifications: Ligand: Anti-BSA Antibody; Particle size (range): 50-175m; Molecular Weight range: 6x104 - 2x107; Matrix: Separopo...
Catalog No. 50-197-814