Learn More
Seracare Life Sciences Inc ANTI-BOVINE IGG (H+L) ANTIBODY
Supplier:  Seracare Life Sciences Inc 52100169
Encompass_Preferred
Affinity purified polyclonal antibody to Bovine IgG, both heavy and light chains (whole IgG) made in Goat. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-668-11