Learn More
Immunology Consultants Monoclonal Mouse anti-Bovine IgM Antibody Clone 2B10, 1.0 mg
SDP
Supplier:  Immunology Consultants MM10A2B10
Encompass_Preferred
This mouse IgG1 is purified using Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with Bovine IgM as determined by ELISA and western blotting. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-241-4298