Learn More
Immunology Consultants anti-Bovine Rotavirus Antibody (Mouse - Clone #100) - Monoclonal
SDP
Catalog No. 5000435288
Encompass_Preferred
This monoclonal antibody provides highly specific detection of bovine rotavirus, a major cause of neonatal diarrhea and economic loss in cattle herds. Clone 100 recognizes a defined viral antigen and delivers strong affinity with minimal nonspecific binding, supporting ELISA, Western blotting, and antigen‑capture assay development. Its clean, reproducible signal enables sensitive detection in fecal samples, culture supernatants, and purified viral preparations. MRTV‑10A‑100 is ideal for veterinary diagnostics, vaccine evaluation, and research on rotavirus epidemiology and pathogenesis. Consistent lot‑to‑lot performance ensures reliable results across diverse assay formats.
Catalog No. 50-004-35288 Supplier Immunology Consultants Supplier No. MRTV10A100
This monoclonal antibody provides highly specific detection of bovine rotavirus, a major cause of neonatal diarrhea and economic loss in cattle herds. Clone 100 recognizes a defined viral antigen and delivers strong affinity with minimal nonspecific binding, supporting ELISA, Western blotting, and antigen‑capture assay development. Its clean, reproducible signal enables sensitive detection in fecal samples, culture supernatants, and purified viral preparations. MRTV‑10A‑100 is ideal for veterinary diagnostics, vaccine evaluation, and research on rotavirus epidemiology and pathogenesis. Consistent lot‑to‑lot performance ensures reliable results across diverse assay formats.